Erythromycin ≥98%
New Product
Erythromycin ≥98%
New Product
Erythromycin ≥98%
Usage: Biochemical/life science applications
 :  (-)-Erythromycin
  •  :114-07-8
  •  :C₃₇H₆₇NO₁₃
  •  :733,93 g/mol
Product Family Options
  • Type de conditionnement
  • Bottle
    1 * 500 mg
    1 * 500 mg - 67.60 SFr.
    67.60 SFr.
    1 * 500 mg
       67.60 SFr.
    New Product
    New Product
  • Bottle
    1 * 1 g
    1 * 1 g - 81.40 SFr.
    81.40 SFr.
    1 * 1 g
       81.40 SFr.
    New Product
    New Product
  • Bottle
    1 * 5 g
    1 * 5 g - 149.00 SFr.
    149.00 SFr.
    1 * 5 g
       149.00 SFr.