Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

Sodium caprylate 99%

 
New Product
Sodium caprylate 99%
New Product
 
Sodium caprylate 99%

 

 

Usage: Organic building block/ reagent.

 :  Octanoic acid sodium acid, Sodium caprylate, Caprylic acid sodium acid, Sodium n-Octanoate
  •  :1984-06-1
  •  :C₈H₁₅O₂·Na
  •  :MFCD00058511
  •  :166,19326 g/mol
Product Family Options

 

  • Type de conditionnement
     
     
  • Bottle
       
       
    1 * 100 g
    1 * 100 g - SFr. 29.50
    SFr. 29.50
    1 * 100 g
       SFr. 29.50

     
    New Product
    New Product
  • Bottle
       
       
    1 * 1 kg
    1 * 1 kg - SFr. 132.00
    SFr. 132.00
    1 * 1 kg
       SFr. 132.00

     
    New Product
    New Product
  • Bottle
       
       
    1 * 500 g
    1 * 500 g - SFr. 46.80
    SFr. 46.80
    1 * 500 g
       SFr. 46.80

     
    New Product
    New Product