Wright Stain 97%
New Product
Supplier:   Apollo Scientific
Wright Stain 97%
Supplier:  Apollo Scientific
New Product
Certificates
Wright Stain 97%
5 Options Available Below
Product Details & Documents
Usage: Organic building block/reagent.
Synonyms:  Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
  • CAS Number:68988-92-1
Product Family Options
Product Information
  • Type de conditionnement
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 26.00 SFr.
    26.00 SFr.
    1 * 5 g
       26.00 SFr.
    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 29.50 SFr.
    29.50 SFr.
    1 * 25 g
       29.50 SFr.
    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 78.00 SFr.
    78.00 SFr.
    1 * 100 g
       78.00 SFr.
    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 300.00 SFr.
    300.00 SFr.
    1 * 500 g
       300.00 SFr.
    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 1 200.00 SFr.
    1 200.00 SFr.
    1 * 2,5 kg
       1 200.00 SFr.
    Specifications:
    New Product
    New Product