MF 37 Modular Ice Machine
Machine à glaçons modulaire, 230 V, 50 Hz, 1 ph, 560×533×525 mm, 215 kg
N° de catalogue SCOMMF37WSOX
New Product
Fournisseur: SCOTSMAN
Aller à
Product Details & Documents
Produces Flake Ice, extruded at a temperature just below 0 °C, with 25% residual water content, hence very rich in moisture.
Resistant stainless steel exterior
AISI304 Stainless steel evaporator and ice scraper blade
Removable side panels on all four sides to aid routine maintenance and servicing
Electronic control board with self diagnostic and malfunction alarms
XSafe: natural sanitation system, integrated inside the ice maker and operating automatically 24/7 to keep equipment clean and safe.
Spécifications
- Poids:215 kg
- Désignation:Machine à glaçons modulaire, 230 V, 50 Hz, 1 ph
- l×p×h:560×533×525 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



