Software SWMP
Logiciel de communication entre l'équipement et le PC, SWMP, Visualisation et impression des procédés réalisés
Catalog # ESPISWMP
New Product
Supplier: RAYPA
Jump to:
Product Details & Documents
Communication software between the equipment and the PC that allows the display and recording in real time or after each cycle performed in the media preparator.
Included items: ethernet cable 1,8 m in length, ethernet to us adapter and USB memory stick with the software
- Allows in-depth analysis of data related to each cycle
- Intuitive display of information on the cycles performed: batch record, outcome, probes temperature, pressure, serial number, autoclave model, errors, etc
- Access is protected by username and password
- Available languages are: English, Spanish, French and Italian
- The export of memory cycles is encrypted (.DAT files)
Specifications
- Désignation:Logiciel de communication entre l'équipement et le PC, SWMP
- Pour:Visualisation et impression des procédés réalisés
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..