External Probe Adapter
Adaptateur externe permettant l'accès à une sonde externe CAP-MP (Ø 3 - 5 mm) pour les processus de validation en continu.
Catalog # ESPICAP-MP
Supplier: RAYPA
Jump to:
Product Details & Documents
Allows access for an external probe to obtain a temperature reading from the culture medium, independent of the reading taken by the equipment's flexible probe.
The port is located on the door of the media preparator
- The TP-VAL-MP temperature probe, compatible for this port, is available upon request
- The external probe adapter is composed of three parts: stainless steel gland, cylindrical chrome-plated brass extension and polypropylene GL-32 cap
Specifications
- Désignation:Adaptateur externe permettant l'accès à une sonde externe CAP-MP (Ø 3 - 5 mm) pour les processus de validation en continu.