Validation and Qualification Sets
Product Details & Documents
They make it possible to obtain an independent reading of the culture medium temperature, separate from the one collected by the equipment’s flexible probe. Essential for the calibration and/or qualification of media preparators.
Recommended for distributors or service providers specialised in sterilisation equipment
Ideal for customers with their own technical service team who will handle the qualification and maintenance throughout the equipment’s lifespan
In the validation process, it is crucial to have options that guarantee the proper calibration and qualification of the equipment. For this purpose, we offer two specific kits to address these needs: calibration set and qualification set.
Calibration set: Calibration of media preparators can be performed with the TP-VALMP calibration set, allowing for the adjustment of the equipment temperature probes to ensure accurate measurements against a known standard. This guarantees that the equipment operates within specified temperature limits.
Qualification set: To ensure that media preparators function according to design specifications, we offer three specialised qualification sets based on the selected model: TP-VAL-MP-20, TP-VAL-MP-40/60, and TP-VALMP-80/100. This process includes verification and documentation certifying that the equipment meets expected technical and performance standards. It is suitable for conducting both Installation Qualification (IQ) and Operational Qualification (OQ).
Bestellinformationen: In order to use the calibration set, it is necessary to purchase the CAP-MP external probe adapter accessory.
Spécifications
- Désignation:Kit d'étalonnage compatible avec les préparateurs de milieux Classic et Top Line TP-VAL-MP, pour le réglage des sondes de température de l'équipement afin de garantir des mesures précises par rapport à une norme connue.
- Seq:1
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..