Flexible Temperature Probe
Flexible Temperature Probe
Sonde de température flexible PT-100 classe A, PT-2-B-AH, 23 kg, , Surveillance de la stérilisation des charges liquides. Compatible avec les autoclaves de la série AHS-B
N° de catalogue ESPIPT-2-B-AH
Fournisseur:   RAYPA
Flexible Temperature Probe
N° de catalogue ESPIPT-2-B-AH
Fournisseur:  RAYPA
Flexible Temperature Probe
Flexible Temperature Probe
Sonde de température flexible PT-100 classe A, PT-2-B-AH, 23 kg, , Surveillance de la stérilisation des charges liquides. Compatible avec les autoclaves de la série AHS-B
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
PT-100 flexible probe recommended for monitoring the sterilisation of liquid loads.
  • 120  mm flexible PT-100 Class A temperature probe
  • Probe calibration via autoclave microprocessor (admin access only)
  • Data analysis available through SW8000 software
  • Enables sterilisation control based on load temperature
After installing this accessory, the temperature regulation of the sterilisation cycle can either be controlled by the main chamber temperature sensor or both the main chamber temperature sensor and the temperature sensor of the flexible temperature probe.
The temperature control by the flexible temperature probe is especially advantageous for processes involving the sterilisation of large volumes of liquids, where the sterilisation process is regulated by both the temperature achieved in the center of the liquid sample as well as the temperature achieved in the sterilisation chamber. Furthermore, should the autoclave be opened at chamber temperatures higher than 80ºC there is a risk of liquids boiling over which can be avoided if the temperature of the sample is controlled throughout the sterilisation procedure.
Attention: Factory installation required.
Spécifications
  • Désignation:
    Sonde de température flexible PT-100 classe A, PT-2-B-AH, 23 kg
  • Pour:
    Surveillance de la stérilisation des charges liquides. Compatible avec les autoclaves de la série AHS-B
  • Cat. No.:
    ESPIPT-2-B-AH