Dextran sulphate
Fournisseur: G-Biosciences
Aller à
À propos de cet article
Dextran sulfate sodium (CAS 9011‑18‑1) is a versatile high‑molecular‑weight polyanion, essential in molecular biology (e.g., hybridisation), biochemistry (e.g., protein/lipoprotein precipitation), virology, inflammation modeling, and nanomedicine construction
- White to off-white crystalline powder
- BSE/TSE Free
- Loss on drying: ≤10%
- Free Sulfate: ≤0,5 %
- pH (1%, Water) @ 25C: 5,0 to 7,5
- Chloride: ≤1000 ppm
- Sulfur Content: 17,0 to 20,0%
Synonymes: Dextran sulphate sodium from Dextran
- Numéro CAS:9011-18-1
- Poids moléculaire:>500,000 g/mol
2 variantes
Product Details & Documents
Dextran sulfate sodium (CAS 9011‑18‑1) is a versatile high‑molecular‑weight polyanion, essential in molecular biology (e.g., hybridisation), biochemistry (e.g., protein/lipoprotein precipitation), virology, inflammation modeling, and nanomedicine construction
- White to off-white crystalline powder
- BSE/TSE Free
- Loss on drying: ≤10%
- Free Sulfate: ≤0,5 %
- pH (1%, Water) @ 25C: 5,0 to 7,5
- Chloride: ≤1000 ppm
- Sulfur Content: 17,0 to 20,0%
Synonymes: Dextran sulphate sodium from Dextran
- Numéro CAS:9011-18-1
- Poids moléculaire:>500,000 g/mol



