Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

Prodigiosin ≥95% apoptotic agent

Marketsource
Fournisseur:   Abcam
Prodigiosin ≥95% apoptotic agent
Fournisseur:  Abcam
Marketsource
Certificats

2 variantes

Product Details & Documents

Potent apoptotic agent. Antibiotic agent. Induces caspase-dependent apoptosis. Inhibits mTORC1 and mTORC2. Reactivates p53. Inhibits iNOS expression. Shows anti-ischemic and antimalarial effects in vivo.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • Poids moléculaire:323,4 g/mol
Spécifications de la famille de produits

Résultats des tests de spécification

  • Constituents::
    100% Prodigiosin
Product Family Options

Informations sur le produit

  • Disponibilité
    Prix
  • Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 mg
    1 * 1 mg - 444.00 SFr.
    444.00 SFr.
    1 * 1 mg
       444.00 SFr.

    Spécifications :
    Marketsource
    Marketsource
  • Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 250 µG
    1 * 250 µG - 150.00 SFr.
    150.00 SFr.
    1 * 250 µG
       150.00 SFr.

    Spécifications :
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.