Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

Thymidine (2'-deoxythymidine) ≥98% Pyrimidine deoxynucleoside

Fournisseur:   Abcam
Thymidine (2'-deoxythymidine) ≥98% Pyrimidine deoxynucleoside
Fournisseur:  Abcam
Certificats

2 variantes

Product Details & Documents

Pyrimidine deoxynucleoside. Potent thymidine kinase substrate (IC₅₀ = 16 nM). Pairs with deoxyadenosine in double-stranded DNA. Synchronises cells in S phase in vivo.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymes:  1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine
  • Numéro CAS:146183-25-7
  • Poids moléculaire:242,23 g/mol
Spécifications de la famille de produits

Résultats des tests de spécification

  • Constituents::
    98% Thymidine
Product Family Options

Informations sur le produit

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.