Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Baicalein ≥98% Lipoxygenase inhibitor

Baicalein ≥98% Lipoxygenase inhibitor

N° de catalogue ABCAAB120723-50
Marketsource
Fournisseur:   Abcam
Baicalein ≥98% Lipoxygenase inhibitor
N° de catalogue ABCAAB120723-50
Fournisseur:  Abcam
Marketsource
Numéro CAS:  491-67-8
Baicalein ≥98% Lipoxygenase inhibitor
Numéro CAS:  491-67-8
Prix
SFr. 94.80
Connexion Pour la disponibilité
1 * 50 mg
Product Details & Documents

Cell-permeable flavonoid. Lipoxygenase inhibitor (IC₅₀ values are 0,64 and 1,6 μM at 12- and 15-lipoxygenases, respectively). Shows antioxidant and anti-inflammatory properties. Also induces apoptosis. Additionally, inhibits α-syn oligomerisation, β-Amyloid induced neurotoxicity and amyloid protein aggregation.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymes:  5,6,7-Trihydroxyflavone, 5,6,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 5,6,7-Trihydroxy-2-phenylchromen-4-one
  • Numéro CAS:491-67-8
  • Poids moléculaire:270,24 g/mol
Spécifications

Résultats des tests de spécification

  • Constituents::
    100% Baicalein
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.