Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Opti-Klear™ Live Cell Imaging Buffer 5X, Abcam

Opti-Klear™ Live Cell Imaging Buffer 5X, Abcam

Opti-Klear™ Live Cell Imaging Buffer 5X
N° de catalogue ABCAAB275939-30
Fournisseur:   Abcam
Opti-Klear™ Live Cell Imaging Buffer 5X, Abcam
N° de catalogue ABCAAB275939-30
Fournisseur:  Abcam
Numéro CAS:  
Opti-Klear™ Live Cell Imaging Buffer 5X, Abcam

Opti-Klear™ Live Cell Imaging Buffer 5X

Numéro CAS:  
Prix
SFr. 125.00
Connexion Pour la disponibilité
1 * 30 mL
Product Details & Documents

Hepes based imaging buffer formulated to lower background fluorescence and maintain proper pH and osmolarity for hours at 37°C. ab275939 provides an energy source and preserves fluorescent signals.

  • Constituents: 99% HEPES

Attention: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Spécifications

Spécifications

  • Désignation:
    Opti-Klear™ Live Cell Imaging Buffer 5X
  • Cat. No.:
    ABCAAB275939-30
  • Température de stockage:
    Store at +4°C.

Spécifications