Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Raccords et connecteurs
Vent Assembly, 2 Port, Straight
Vent Assembly, 2 Port, Straight
Vent Assembly, 2 Port, Straight
DISI4050-0065
:
DISTEK
New Product
Vent Assembly, 2 Port, Straight
DISI4050-0065
:
DISTEK
New Product
Filetage:
GL-45
Désignation:
Vent assembly, 2-port, straight, universal, straight, GL 45, external hose barb: 6,35 mm
Matière:
Acier inoxydable 316