Sampling and Mailing Tubes with Screw Cap, PP
SEMA11543
: SEMADENI
New Product
- Capacité:180 ml
- Couleur:transparent
- Désignation:Container with screw cap, Ø ext.×H: 63×80 mm
- Seq:2
Transparent tubes, with flat base and tightly closing screw caps.
For universal use
Suited for collecting and transporting samples in liquid or solid form