Specifications
- Emballage:6 cans per box, 1000 ml each
- Désignation:Premium liquid soap, refill
Specifications
About this item
This soap has a dermatologically tested formula, skin-friendly pH and fat-dissolving ingredients, meaning it is suitable for food preparation and washing greasy hands.
Save staff time with effortless refilling – certified by the Swedish Rheumatism Association, tested according to the Design for All test, published in the Swedish Design Research Journal, 2011
- Help secure good hand hygiene - the factory-sealed bottle with a new pump for every refill helps reduce the risk of contamination and protect the formulations all the way to the hands
- Reduce waste - the bottle is collapsible, leading to 70% less waste volume (based on Essity test)
-
Perfume free fragrance, premium quality
Colour: Transparent
Compatible with S4 dispensers
To remove dirt, oil and grease, Tork oil and grease liquid soap is formulated with a fat-dissolving ingredient that is highly effective yet gentle on the skin.
No fragrance or colour is added, so it is gentle to the skin. It is suitable for Tork S4 dispensers, which are certified easy to use and provide good hand hygiene to all users.
Nordic Swan Ecolabel, 2090 0085.