Spécifications
- Capacité:1000 ml
- Couleur:blanc
- Désignation:Clarity foam soap starter pack with dispenser, refill
- l×p×h:114×113×292 mm
Spécifications
À propos de cet article
Tork clarity foam soap starter pack with white dispenser S4, sustainable formula, dispenser.
Establish great hand hygiene with this starter pack, which includes a Tork S4 skincare dispenser and a Tork clarity foam soap refill
- Save your staff time, with certified effortless maintenance and intuitive refilling in less than 10 seconds
- Limit the risk of contamination - the factory-sealed bottle comes with a new pump for every hand soap refill
-
Improve your eco credentials, as this formula is readily biodegradable with 99% of the ingredients from natural origins
Elevated design
Suitable for S4 refills from Tork
Improve hand hygiene and cut waste with our Tork Clarity Foam Soap Starter Pack. The soap lowers water consumption by 35% and the bottle collapses as the contents are used. Also to improve sustainability, ingredients are chosen for their low impact on aquatic life, with 99% of them from natural origins.
The soap comes with a dispenser that was made for easy use – it has an effortless maintenance certification and takes less than 10 seconds to refill.
Based on an Internal panel test comparing with Tork mild foam soap
Based upon the EU Ecolabel, CDV (Critical Dilution Volume) related to Toxicity to aquatic organisms is below specified limits and is two times lower than the TORK Mild foam soap formula
Based on internal panel test soap refilling time
EU Ecolabel, SE/030/002.