Tork W1 Heavy-Duty Cleaning Cloths
Catalog # SCAA510204
Supplier: Essity
Specifications
- Couleur:bleu
- Désignation:Cloths, refill, premium, compatible with Tork W1 dispensers, 1×1000 cloths
- Longueur:380 mm (Sheet length)
- L×l:380000×420 mm (Roll size)
Specifications
About this item
Easily carry, open and dispose of packaging. Clean tight spaces effectively with this soft and flexible cleaning cloth.
Use less effort as heavy-duty cloth rapidly absorbs both oils and liquids
Save time when refilling - this high-capacity roll lasts longer
Busy workspaces need multipurpose cleaning cloths that won't fall apart when it needed. Soft yet strong Tork cleaning cloth is ideal for most light cleaning tasks. Less effort is needed as it absorbs oils and liquids easily, helping staff to work efficiently. The longer-lasting, high-capacity roll fits Tork floor or wall stand dispensers. These safe and reliable dispensers are easy to load and offer single-handed tear-off for a boost of hygiene and productivity.
Certifications: FSC MIX Credit, SA-COC-008266.