Culture Media, Ready to Use, in Bags
Catalog # LIOF499070
Supplier: LIOFILCHEM
New Product
CAS Number:
Specifications
- Cdt:3
- Désignation:Buffered NaCl peptone solution pH 7,0
- Volume final:3 L
Specifications
About this item
Microbiology culture media bags, are pre-prepared, sterile liquid culture media contained in plastic bags. They are used in microbiology laboratories for various applications, including food and pharmaceutical microbiology.
Time saving: Since the media is pre-prepared, there is no preparation time required. You can start using the media immediately upon arrival in the lab
- Reduced risk of contamination: Pre-sterilised media reduces the risk of contamination compared to manually prepared media
- Batch consistency: Ordering large batches of RTU media can reduce the number of performance tests required, ensuring consistency between experiments
These bags are designed to be connected to a gravimetric dilutor, simplifying the process of weighing and diluting samples.