Attaches universelles à trois pattes
Katalognr. WITG8.430.000
Anbieter: witeg Labortechnik
Technische Daten
- Largeur de serrage:0 - 120 mm
- Cdt:1
Technische Daten
Über diesen Artikel
These universal three-pronged clamps are made of laquered aluminium, with nickel plated brass screws.
- Jaws are covered with a layer of cork to protect clamped items