Volumetric flasks, class A, brown markings
613-4125
- RN:10/19
- Capacité:15 ml
- Environmentally Preferable:
- Cdt:2
- Tolérance:0,04 ± ml
DURAN® borosilicate glass 3.3 with PE stopper.
- With spout
- Conformity certified
- Delivered with batch number and have enclosed one batch certificate per package