Powder funnels
Powder funnels
150 mm, 138 mm, 29 mm
N° de catalogue LPIT291044
Fournisseur:   LP ITALIANA
Powder funnels
N° de catalogue LPIT291044
Fournisseur:  LP ITALIANA
Powder funnels
Powder funnels
150 mm, 138 mm, 29 mm
Prix
216.00 SFr.
Connexion Pour la disponibilité
1 * 10 ST
Product Details & Documents
Funnels, made of PP, translucent, with short, wide stem. Used for transferring powders. Also suitable for liquids.
  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable
Spécifications
  • Ø ext. entonnoir sup.:
    150 mm
  • Longueur:
    138 mm
  • Ø ext. tige:
    29 mm