Volumetric flasks, class B, white markings
Catalog # HECH42089092
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- RN:-
- Capacité:3000 ml
- Seq:01
- Environmentally Preferable:
- Cdt:1
- Tolérance:1,2 ± ml
Specifications
About this item
Borosilicate glass 3.3.
- Higher accuracy than standard class B, Tolerance: 1,5 times of the official tolerance of class A
- Calibrated to contain (marked 'In')
*Wide neck
Certifications: In accordance with DIN EN ISO 1042