L-Lactate dehydrogenase (from Muscle)
Catalog # CHMCAB1222
Supplier: Chemicon
CAS Number:
Specifications
- Cdt:25 mg
- N ° CAS.:9001-60-9
- Source:Muscle
- Species:
- Environmentally Preferable:
- Conditions de stockage:Refrigerator
- MDL No.:MFCD00131466
- Nom de l'enzyme:L-Lactate dehydrogenase
- Synonymes d'enzyme:LDH