ContactUsLinkComponent
Hand disinfectant gels, Disinfector 70%
Hand disinfectant gels, Disinfector 70%
Catalog # 115-0784
Supplier:  Plum Safety
undefined
Hand disinfectant gels, Disinfector 70%
Catalog # 115-0784
Supplier:  Plum Safety
Specifications
  • Désignation:
    Disinfectant gel 70%, for CombiPlum dispenser, 1000 ml refill
  • Seq:
    02
  • Cdt:
    6
Specifications
About this item
Disinfection gel for bacteria removal of the hands.
  • With reconditioner
  • Approved in accordance with the European Standard pr. EN12054 and EN1500
Ideal for use in the food industry and in other situations where high standards of hygiene are required.
Certifications: EN12054, EN1500