Body lotion, Lindesa®
Catalog # FAWE13640-010
Supplier: FAWECO HAUTSCHUTZ
Specifications
- Désignation:Body lotion, Lindesa®, 50 ml tube
- Seq:01
- Cdt:50 mL
Specifications
About this item
Barrier cream and care cream with beeswax is designed to care for skin exposed to minor contamination, and is also suitable for after-work care.
- Lightly greasy, absorbs quickly
- Special care ingredients improve the skin's elasticity and suppleness
- Does not affect the sense of touch
- Emulsion type: O/W, neutral pH level, silicone-free, scented
The cream strengthens the skin's immunity, prevents skin damage from exposure to external contamination, and has a soothing effect on irritated skin. It smooths and nourishes without any long-lasting, greasy film.
Caution: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904