Ruban adhésif, fixation Transpore™
Catalog # MMMEM1527-1
Supplier: 3M MEDICAL
Specifications
- Désignation:Sparadrap chirurgical, Transpore™
- L×l:9,1 m × 25 mm
- Seq:01
- Cdt:12 Roll
Specifications
About this item
This transparent, perforated plastic tape can be used for different applications, such as securing tubing and devices (e.g., catheters, I.V. tubing) and anchoring bulky dressings.
- Tears easily, works reliably
- Easy to handle with gloves
- Good adhesion to skin and tubing
- Hypoallergenic and not made with natural rubber latex
- Water resistant
- Allows clinician to tailor the tape for the various dressings and devices
- Economical single use rolls