Wash bottles, wide mouth, with coloured caps, labeled, Safety-Vented™
Katalognr. BELAF118320019
Anbieter: Bel-Art Products, a Part of SP
Technische Daten
- Code couleur:naturel
- Capacité:1000 ml
- Seq:02
- Cdt:2
- Impression:Éthanol
Technische Daten
Über diesen Artikel
LDPE, with PP spray cap (vented).
Bottles feature a 53 mm wide mouth for easy refilling. They are pre-printed with the chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment. Leak-proof, these translucent LDPE bottles have color-coded PP closures to aid in quick and easy identification.
A unique valve prevents pressure build-up inside these Right-to-Know safety-labeled wash bottles, eliminating potentially dangerous drips. This valve allows dispensing in an upright or inverted position. Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
Packaging: 500 ml bottles are packaged as 4/bag and 1000 ml bottles are packaged as 2/bag.