Forceps, curved, SP Bel-Art
Catalog # BELAH379400000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Type:Incurvées, dentelées
- Longueur:98 mm
- Cdt:2
Specifications
About this item
Gently curved stainless steel forceps with exceptionally fine tips which are coated with brown Fluo-Kem® PTFE.
- FDA compliant for food contact
- Serrated, curved tips