Autoclaving wraps, Super Polyfoil™, SP Bel-Art
293-2121
- Couleur:bleu
- Désignation:Enveloppe PE d'autoclavage avec protection chimique pour couvrir les récipients et les incubateurs
- Dimensions:130×130 mm
- Cdt:100
Polyester backed foil provides a chemically inert barrier to cover vessels for autoclaving, incubating or storage.
- Thermoplastic layer is laminated
- Folds easily and flattens without kinking
- Resistant to punctures, tears and corrosion
- Polyester film blocks pinholes and resists organic solvents and caustic agents
: Complies with FDA for food contact applications