Weighing dishes, aluminium
Catalog # HEATHS14522
Supplier: HEATHROW
Specifications
- Ø ext.:101 mm
- Hauteur:8 mm
- Cdt:50
Specifications
About this item
Weighing dishes, made from light gauge aluminium are designed for moisture determination balances. Ideal for general-purpose weighing and dispensing.
- Used as evaporating dish or dust cover
- Lightweight, sturdy dish has no coating and is oil free
- Slightly tapered for stacking