Capillary cutter, CleanCut
Coupe-tubes CleanCut, avec lame de rechange
N° de catalogue 590-0850
Fournisseur: S.C.A.T.
Aller à
Product Details & Documents
This cutter is made of PP, blue, with stainless steel blade.
- Separates capillary tubes safely and reliably
- With useful openings for measuring the capillary diameter
- Prevents injuries and ensures a clean cut
The universal tube cutter for HPLC capillaries - the tube guide enables a comfortable, straight cut. With spare blade for exchange.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Spécifications
- Désignation:Coupe-tubes CleanCut, avec lame de rechange