0
0,00 SFr.
Order Entry
Connexion
/
S'inscrire
Products
Services
Digital Solutions
Applications
Support
About
Saisie de commande
Statut de la commande
Votre compte
Panier
Switzerland
Saisie de commande
Statut de la commande
Votre compte
Panier
ContactUsLinkComponent
Accueil
Adaptateurs
Raccords rodés
Connecting adapters, Clear-Seal®, WHEATON®
Connecting adapters, Clear-Seal®, WHEATON®
Connecting adapters, Clear-Seal®, WHEATON®
Catalog # WHEA139479
Supplier:
DWK Life Sciences
Expand image
Connecting adapters, Clear-Seal®, WHEATON®
Catalog # WHEA139479
Supplier:
DWK Life Sciences
Specifications
Rodage femelle:
19/22 NS
Cdt:
1
Specifications
About this item
Product Details
Product Details