ContactUsLinkComponent
Canister for cleanroom wipes, pre-saturated, SATWipes®
Canister for cleanroom wipes, pre-saturated, SATWipes®
Catalog # 115-1874
Supplier:  CONTEC
undefined
Canister for cleanroom wipes, pre-saturated, SATWipes®
Catalog # 115-1874
Supplier:  CONTEC
Specifications
  • Désignation:
    Canister avec couvercle à fermeture par clips
  • Cdt:
    10
  • Pour:
    Rolls, 152×229 mm
Specifications
About this item
This reusable canister holds pre-saturated wipes that are 6" x 9" (152×229 mm) and are dispensed from center-pull rolls.
Canister comes with a snap close lid that helps keep wipes saturated and clean.
Caution: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904