Spectroquant reference standard for Picco, Move and Multy colorimeters, Supelco®
Catalog # 1.19301.0001
Supplier: Merck
CAS Number:
Specifications
- Cdt:1
- Désignation:Spectroquant® reference standard for chlorine, chlorine dioxide and ozone
Specifications
About this item
A reference standard for chlorine, chlorine dioxide and ozone, that is designed to reassure the user about the accuracy and reliability of the results from the Picco and Multy colorimeters.
Delivery: Standard includes four sealed cells including a Zero cell and stable coloured solutions and is supplied in a handy case with quality data, lot number and expiry date.