Pinces pour flacons
Katalognr. 232-4152
Anbieter: USBECK Laborgeräte
Technische Daten
- Longueur:300 mm
- Cdt:1
- Serrage:45 - 70 mm
Technische Daten
Über diesen Artikel
Flask tongs made from stainless steel, with vinyl coated jaws.
Flask tongs made from stainless steel, with vinyl coated jaws.