- Désignation:deconex® SOLARSEPT
- Emballage:500 ml plastic bottle with spray nozzle
- Cdt:1
deconex® SOLARSEPT is used for disinfection when a short exposure and drying time is required.
- aldehyde-free
- Ready-to-use, with rapid effect ime and short drying time
- Can also be sprayed on
- Broad spectrum of activity including tuberculocidal effect
- Pleasant fresh smell-
deconex® SOLARSEPT is used on medical equipment and devices and other small surfaces, such as examination couches, operating tables and adjacent work surfaces, user interfaces on medical equipment such as patient monitors, ultrasound and other examination equipment, furniture in patients' rooms, medical and dental practices, medical laboratories and cosmetic studios.
Wipe disinfection: Pour the product out of the dosing bottle onto a clean cloth or directly onto the surface to be treated (approximately 25 ml product/m²).
Gently wipe the surface all over with steady movements. Makesure that the surface is completely wetted.
Spray disinfection: Spray the surface to be treated, ensuring in particular that it is completely wetted.