Sodium thiosulfate 0.2 mol/l (0.2 N) en solution aqueuse
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7772-98-7
:
Formula:Na₂S₂O₃
MDL Number:MFCD00003499
:158.11 g/mol
Storage Temperature:Température ambiante
EINECS:231-867-5
Related Information
Certificates
Search by Lot Numberspec_VWRC302332.1000
Product specification
VWR® Volumetric Solutions for Titration
Product Literature
Frequently Bought Together