Order Entry
Switzerland
ContactUsLinkComponent
Penicillinase (beta-Lactamase) (from Enterobacter cloacae)
Penicillinase (beta-Lactamase) (from Enterobacter cloacae)
Catalog # CHMCAB1234
Supplier:  Chemicon
CAS Number:  
Penicillinase (beta-Lactamase) (from Enterobacter cloacae)
Catalog # CHMCAB1234
Supplier:  Chemicon
CAS Number:  

Specifications

  • Cdt:
    25 mg
  • N ° CAS.:
    9001-74-5
  • Source:
    Enterobacter cloacae
  • Conditions de stockage:
    Refrigerator
  • Nom de l'enzyme:
    Penicillinase (beta-Lactamase)

Specifications

About this item