Unité de commande, B-436
Unité de commande, B-436
N° de catalogue 458-2230
Fournisseur: BUCHI
Aller à
Product Details & Documents
Control unit for the time and output management (both of which are programmable) of a maximum of two K-435 digestion units with B-414 scrubber.
Ordering information: The B-436 control unit is supplied with a control cable for 1 digestion device, a power cord and the operating manual.
Spécifications
- Désignation:Unité de commande, B-436