ContactUsLinkComponent
Crème de protection des mains, Lindesa K
Crème de protection des mains, Lindesa K
N° de catalogue FAWE13642005
Fournisseur:  FAWECO HAUTSCHUTZ
undefined
Crème de protection des mains, Lindesa K
N° de catalogue FAWE13642005
Fournisseur:  FAWECO HAUTSCHUTZ
Spécifications
  • Désignation:
    Protective hand cream, Lindesa® K with camomile, 50 ml tube
  • Cdt:
    24
Spécifications
À propos de cet article
The cream strengthens the skin's immunity, prevents skin damage from exposure to external contamination, and soothes irritated skin without leaving a greasy film. It also is suitable for the conditioning of stressed skin.
  • Does not leave any grease marks and is quickly absorbed
  • O/w emulsion
  • Silicone-free, perfumed
Ideal for use in workplaces where no greasy residue can be left on surfaces, such as when varnishing, in the production of electronics, or during final inspection and testing.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904