ContactUsLinkComponent
Crème de protection des mains, Lindesa® F
Crème de protection des mains, Lindesa® F
  FAWE1007167
undefined
Crème de protection des mains, Lindesa® F
  FAWE1007167
  • Désignation:
    Protective hand cream, Lindesa® F, 100 ml tube
  • Cdt:
    100 mL
Suitable when handling water-soluble working materials like aqueous solutions (acids, alkaline, or saline solutions), alcohols, drilling emulsions, and cooling lubricants as well as when working with cement, lime, or other similar materials. Also suitable for the caring of strongly degreased skin after work..
  • W/o emulsion
  • Greasy cream
  • Silicone-free, perfumed, contains fats
 : *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904