HPASMC, Cryopreserved
HPASMC, Cryopreserved
HPASMC, cryopreserved, 0,5M cells (5×10⁵ cells/vial), Cryopreserved, Single donor, 1 Vial
N° de catalogue PRTV1210613
Fournisseur:   PROVITRO
HPASMC, Cryopreserved
N° de catalogue PRTV1210613
Fournisseur:  PROVITRO
Numéro CAS:  
HPASMC, Cryopreserved
HPASMC, Cryopreserved
HPASMC, cryopreserved, 0,5M cells (5×10⁵ cells/vial), Cryopreserved, Single donor, 1 Vial
Numéro CAS:  
Prix
SFr. 1’160.00
Connexion Pour la disponibilité
1 * 1 Vial
Product Details & Documents
HPASMC - Human pulmonary artery smooth muscle cells - are obtained from human tissue, in particularly from the pulmonary artery , initially isolated in their in vivo state. The cells are then transitioned into a primary cell culture and subsequently aliquoted and frozen for preservation. It's crucial to note that HPASMC cultures undergo careful methods during isolation, ensuring that they remain untransformed and unmutated, maintaining characteristics reflective of their in vivo state. These primary cultures have a limited in-vitro lifespan.
  • It's crucial to note that HPASMC cultures undergo careful methods during isolation, ensuring that they remain untransformed and unmutated, maintaining characteristics reflective of their in vivo state. These primary cultures have a limited in-vitro lifespan.
  • Each batch of HPASMC undergoes rigorous testing, specifically assessing their proliferative capacity. These assessments are conducted using provitro's culture reagents, as detailed in the accompanying certificate of analysis. This stringent testing process ensures the consistency and reliability of HPASMC cultures for research and experimental applications
Each batch of HPASMC undergoes rigorous testing, specifically assessing their proliferative capacity. These assessments are conducted using provitro's culture reagents, as detailed in the accompanying certificate of analysis. This stringent testing process ensures the consistency and reliability of HPASMC cultures for research and experimental applications. This product contain 500,000 cells/cryovial.
Storge: Check the cryovial for signs of damage upon receipt. Since the cryopreserved HPASMC samples are shipped on dry ice, they must be immediately transferred to a freezer at −80 °C for storage, either for a period of up to one week or after the thawing process described below. In cases where longer storage is required, the vial should be transferred directly to liquid nitrogen at a temperature of −196 °C. This ensures optimal preservation of the vial. This ensures optimal preservation of the cryopreserved HPASMC samples for longer-term storage requirements.
Storge: Check the cryovial for signs of damage upon receipt. Since the cryopreserved HPASMC samples are shipped on dry ice, they must be immediately transferred to a freezer at −80 °C for storage, either for a period of up to one week or after the thawing process described below. In cases where longer storage is required, the vial should be transferred directly to liquid nitrogen at a temperature of −196 °C. This ensures optimal preservation of the vial. This ensures optimal preservation of the cryopreserved HPASMC samples for longer-term storage requirements.
Spécifications
Spécifications
  • Format de cellule:
    Cryopreserved
  • Cdt:
    1 Vial
  • Donneur unique/donneurs regroupés:
    Single donor
  • Désignation:
    HPASMC, cryopreserved, 0,5M cells (5×10⁵ cells/vial)
Spécifications
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..