Stainless Chain
Stainless steel chain, 3,2 Kg, Acier inoxydable 316, 50 m
N° de catalogue SAMP5070S-50
Fournisseur: Sampling Systems
Aller à
Product Details & Documents
Our popular range of accessories are essential when sampling with Sub Samplers (Bomb Sampler) and bailers. Stainless chain provides great chemical resistance.
Spécifications
- Désignation:Stainless steel chain, 3,2 Kg
- Longueur:50 m
- Matière:Acier inoxydable 316