MAST® ASSURE Vibrio Agglutinating Antiseras
Catalog # MAST2215001
Anbieter: MAST DIAGNOSTICA
New Product
CAS-Nummer:
Technische Daten
- Cdt:2 mL
- Désignation:Vibrio cholerae Bengal O139
- Seq:01
Technische Daten
Über diesen Artikel
An extensive range containing both polyvalent and monovalent diagnostic antisera for the identification of Vibrio spp according to the expression of somatic antigens. Bacterial serotype can be determined by both slide agglutination and test tube agglutination methods.
- High quality performance
- Simple slide and test tube agglutination methods
- Easy to read results
Liquid stable antisera for the determination of O antigens for the serological identification of Vibrio cholerae.