High Vis Green Stretch Trousers, 2647 GSTP
Supplier: FRISTADS KANSAS
New Product
Certificates
About this item
Sustainable high-vis stretch trousers ideal for service and logistics. Full 4-way stretch with rib-knit stretch panels at the waist for double the comfort. Made of bio-based and recycled materials and reinforced in exposed areas.
- Concealed front button
- Stiby concept, large bellowed leg pocket with snap fastening and zip pocket
- Durable ripstop 4-way stretch at seat and back of thighs
- D-ring with two front pockets and two bellowed back pockets, one with zip
- Double reinforced crotch seam
- CORDURA®-reinforced knee pockets with inside opening with height adjustment for knee pads in knee pocket
- Hidden boot hook
Material: Stretch fabric 44% PTT (partly bio-based material), 44% polyester, 12% recycled polyester. Ripstop stretch fabric 87% polyamide, 13% elastane.
Weight: Stretch fabric 260 g/m². Ripstop stretch fabric 260 g/m².
Certifications: ISO 20471 class 1, EN 14404, Environmental product declaration (EPD).
Specifications
- Catalog No:
- KWIN134243171C54
- KWIN134243171C52
- KWIN134243171C50
- KWIN134243171C48
- KWIN134243171C46
- KWIN134243171C66
- KWIN134243171C44
- KWIN134243171C64
- KWIN134243171C62
- KWIN134243171C60
- KWIN134243171C58
- KWIN134243171C56
- Modèle:
- 2647 GSTP
- 2647 GSTP
- 2647 GSTP
- 2647 GSTP
- 2647 GSTP
- 2647 GSTP
- 2647 GSTP
- 2647 GSTP
- 2647 GSTP
- 2647 GSTP
- 2647 GSTP
- 2647 GSTP
- Applications des tenues:
- Services
- Services
- Services
- Services
- Services
- Services
- Services
- Services
- Services
- Services
- Services
- Services
- Genre:
- Pour hommes
- Pour hommes
- Pour hommes
- Pour hommes
- Pour hommes
- Pour hommes
- Pour hommes
- Pour hommes
- Pour hommes
- Pour hommes
- Pour hommes
- Pour hommes
- Longueur de manches ou de jambes:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Tissu:
- Stretch fabric 44% PTT (partly bio-based material), 44% polyester, 12% recycled polyester. Ripstop stretch fabric 87% polyamide, 13% elastane.
- Stretch fabric 44% PTT (partly bio-based material), 44% polyester, 12% recycled polyester. Ripstop stretch fabric 87% polyamide, 13% elastane.
- Stretch fabric 44% PTT (partly bio-based material), 44% polyester, 12% recycled polyester. Ripstop stretch fabric 87% polyamide, 13% elastane.
- Stretch fabric 44% PTT (partly bio-based material), 44% polyester, 12% recycled polyester. Ripstop stretch fabric 87% polyamide, 13% elastane.
- Stretch fabric 44% PTT (partly bio-based material), 44% polyester, 12% recycled polyester. Ripstop stretch fabric 87% polyamide, 13% elastane.
- Stretch fabric 44% PTT (partly bio-based material), 44% polyester, 12% recycled polyester. Ripstop stretch fabric 87% polyamide, 13% elastane.
- Stretch fabric 44% PTT (partly bio-based material), 44% polyester, 12% recycled polyester. Ripstop stretch fabric 87% polyamide, 13% elastane.
- Stretch fabric 44% PTT (partly bio-based material), 44% polyester, 12% recycled polyester. Ripstop stretch fabric 87% polyamide, 13% elastane.
- Stretch fabric 44% PTT (partly bio-based material), 44% polyester, 12% recycled polyester. Ripstop stretch fabric 87% polyamide, 13% elastane.
- Stretch fabric 44% PTT (partly bio-based material), 44% polyester, 12% recycled polyester. Ripstop stretch fabric 87% polyamide, 13% elastane.
- Stretch fabric 44% PTT (partly bio-based material), 44% polyester, 12% recycled polyester. Ripstop stretch fabric 87% polyamide, 13% elastane.
- Stretch fabric 44% PTT (partly bio-based material), 44% polyester, 12% recycled polyester. Ripstop stretch fabric 87% polyamide, 13% elastane.