VAI® STER-AHOL® Wipes
VELTVEL612X12S2320
New Product
- Désignation:VAI® STER-AHOL® wipes
- L×l:304×304 mm
- Cdt:100
STER-AHOL® Wipes may be used as a sanitiser in classified manufacturing and testing environments.
- 100% continuous knit polyester filament is specifically designed to be clean and absorbent
- Gamma irradiated at 10⁻⁶ SAL
- Completetly traceable from start to finish
- Individually bagged with two liner bags
STER-AHOL® Wipes are saturated with denatured ethanol formulated to 70%, with a small percentage of methyl alcohol and 30% USP water for injection. These wipes can be used in both aseptic and non-aseptic wipe downs on any critical surface that requires cleaning.