Overboots, Antistatic
Catalog # SSSA000607-W-43-48
Supplier: 3S ARBEITSSCHUTZ
New Product
Specifications
- Pointure:43 - 48
- Couleur:blanc
- Cdt:600
Specifications
About this item
Tyvek is a lightweight material with high tear and abrasion resistance. It consists of fine, spun continuous polyethylene fibres that are compressed into a fleece-like material.
- Non-slip sole
- Height: ~570 mm
The air and water vapor permeability promises good wearing comfort and the material offers high protection against airborne particles and numerous water-soluble inorganic chemicals.