Cy5 Dye qPCR Calibration Solution *10000X*
Référence AATB67033
Fournisseur: AAT Bioquest
New Product
Numéro CAS:
Spécifications
- Cdt:100 µl
- Désignation:Cy5 Dye qPCR calibration solution *10000X*
Spécifications
À propos de cet article
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
- Stable for 12 months
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
Attention: For research use only.