SYBR Dye qPCR Calibration Solution *10000X*
Référence AATB67035
Fournisseur: AAT Bioquest
New Product
Numéro CAS:
Spécifications
- Cdt:100 µl
- Désignation:SYBR dye qPCR calibration solution *10000X*
Spécifications
À propos de cet article
SYBR dye qPCR Calibration solution supplied as a 10000X concentrate is compatible with a wide variety of qPCR instruments, including the ABI7500 Fast, QuantStudio™, and ViiA™ seven systems.
- Stable for 12 months
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
Attention: For research use only.